Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins & Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Manuals & Protocols
  • Product FAQs
    Search button Close
            • Order Status
            • Quick Order
            • Sign in
              Sign in
              Don't have an account ? Create Account
              • Account
              • Check Order Status
              • Aspire Member Program
              • Connect: Lab, Data, Apps
              • Custom Products & Projects
              • Services Central

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • BRUCE Antibodies

            Bethyl Laboratories

            BIRC6/Apollon Polyclonal Antibody

            View all (9) BRUCE antibodies
            Datasheet
            Protocols
            Questions & Answers
            Datasheet
            Protocols
            Questions & Answers

            Cite BIRC6/Apollon Polyclonal Antibody

            • Antibody Testing Data (3)
            Application
            BIRC6/Apollon Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            Group 53 Created with Sketch.

            FIGURE: 1 / 3

            BIRC6/Apollon Antibody (A300-367A) in IHC (P)

            Detection of human BIRC6/Apollon by immunohistochemistry. Sample: FFPE section of human breast carcinoma. Antibody: Affinity purified rabbit anti-BIRC6/Apollon (Product # A300-367A) (Lot 2) used at a dilution of 1:5,000 (0.2 µg/mL). Detection: DAB.
            View Product

            BIRC6/Apollon Antibody in Immunohistochemistry (Paraffin) (IHC (P))
            BIRC6/Apollon Antibody in Western Blot (WB)
            BIRC6/Apollon Antibody in Immunoprecipitation (IP)

            Product Details

            A300-367A

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1:2,000-1:10,000
            -

            Immunohistochemistry (IHC)

            1:2,000-1:10,000
            -

            Immunohistochemistry (Paraffin) (IHC (P))

            1:2,000-1:10,000
            -

            Immunoprecipitation (IP)

            2-10 µg/mg lysate
            -
            Product Specifications

            Species Reactivity

            Human, Mouse

            Host/Isotype

            Rabbit / IgG

            Class

            Polyclonal

            Type

            Antibody

            Immunogen

            Region between residue 4775 and the C-terminus (residue 4829) of human Baculoviral IAP repeat-containing protein 6.
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            1 mg/mL

            Purification

            Antigen affinity chromatography

            Storage buffer

            phosphate/tris citrate, pH 7-8

            Contains

            0.09% sodium azide

            Storage conditions

            4°C

            Shipping conditions

            Ambient (domestic); Wet ice (international)

            Product Specific Information

            The recommended shelf life for this product is 1 year from date of receipt.

            Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.

            Target Information

            Apoptosis, or programmed cell death, is related to many diseases, such as cancer. Apoptosis is triggered by a variety of stimuli including members in the TNF family and can be prevented by the inhibitor of apoptosis (IAP) proteins. IAP proteins form a conserved gene family that binds to and inhibits cell death proteases. BRUCE, also known as BIRC6, is an IAP family member protein with a BIR (baculoviral inhibition of apoptosis protein repeat) domain and a UBCc (ubiquitin-conjugating enzyme E2, catalytic) domain. BRUCE regulates p53 and the mitochondrial pathway of apoptosis by facilitating the degradation of apoptotic proteins such as Caspase-9 and SMAC by ubiquitination.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            References (0)

            Have you cited this product in a publication?
            Let us know so we can reference it here.
            Cite this product

            Bioinformatics

            Protein Aliases: APOLLON; baculoviral IAP repeat-containing 6; Baculoviral IAP repeat-containing protein 6; baculovirus inhibitor of apoptosis repeat containing ubiquitin-conjugating enzyme; baculovirus inhibitor of apoptosis repeat containing ubiquitin-conjugating enzyme; accccbaculoviral IAP repeat-containing 6; BIR repeat-containing ubiquitin-conjugating enzyme; BRUCE; Dual E2 ubiquitin-conjugating enzyme/E3 ubiquitin-protein ligase BIRC6; Ubiquitin-conjugating BIR domain enzyme apollon; ubiquitin-conjugating BIR-domain enzyme apollon

            View more View less

            Gene Aliases: A430032G04Rik; A430040A19Rik; AA501170; APOLLON; BIRC6; BRUCE; D630005A10Rik; KIAA1289; mKIAA1289

            View more View less

            UniProt ID: (Human) Q9NR09, (Mouse) O88738

            View more View less

            Entrez Gene ID: (Human) 57448, (Mouse) 12211

            View more View less

            Function(s)

            ubiquitin-protein transferase activity cysteine-type endopeptidase inhibitor activity protein binding ligase activity ubiquitin conjugating enzyme activity UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity ribosomal S6-glutamic acid ligase activity peptidase inhibitor activity coenzyme F420-0 gamma-glutamyl ligase activity coenzyme F420-2 alpha-glutamyl ligase activity protein-glycine ligase activity protein-glycine ligase activity, initiating protein-glycine ligase activity, elongating tubulin-glycine ligase activity protein-glutamic acid ligase activity tubulin-glutamic acid ligase activity

            Process(es)

            protein phosphorylation apoptotic process mitotic nuclear division positive regulation of cell proliferation negative regulation of endopeptidase activity protein ubiquitination regulation of cytokinesis regulation of cell proliferation negative regulation of apoptotic process cell division labyrinthine layer development spongiotrophoblast layer development negative regulation of extrinsic apoptotic signaling pathway placenta development cell cycle regulation of signal transduction negative regulation of peptidase activity
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us
            • Careers
            • Investors
            • News
            • Social Responsibility
            • Trademarks
            • Consumer Health Data Privacy Policy
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Information Center
            • Price & Freight Policy
            • Cookie Preferences

              Your choices regarding cookies on this site

              We and our affiliates and vendors use cookies and similar technologies to operate our sites, recognize visitors to our sites, provide secure log-in, collect statistics to optimize site functionality, and deliver content tailored to your interests. Click Accept All to accept all cookies and go directly to the site, click Reject All to reject all but cookies strictly necessary to the functioning of this site (required cookies), or click on Manage Settings to see detailed descriptions of the types of cookies and choose whether to accept certain cookies while on the site.
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            United States flag icon
            United States

            Your items have has been added!


            Host server : magellan-search-green-56756dfb8b-mfpfg:80/100.66.79.199:80.
            git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.31.1-2025.06.25-1.0