Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
This antibody reacts with mouse SLP65 / BLNK, a cytosolic adaptor protein identified as two phosphoproteins migrating at 68 and 70 kDa in SDS/PAGE (alternatively spliced forms of human SLP65).
Western Blot: Reducing conditions.
B cell linker protein (Blnk) is a protein encoded by the Blnk gene located on chromosome 10 in humans. It is also known as SLP 65 (Src homology 2 domain-containing leukocyte protein of 65 kDa). It derives its name from being an adaptor protein in the B cell receptor signaling cascade with several supporting studies showing improper development of B cells in the absence of Blnk. Blnk undergoes tyrosine phosphorylation by Syk, a B cell receptor activated kinase, which provides a platform for interaction of signaling intermediaries such as the Vav GEF and Grb2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B cell adaptor containing SH2 domain; B cell linker protein; B-cell activation; B-cell adapter containing a SH2 domain protein; B-cell adapter containing a Src homology 2 domain protein; B-cell linker protein; Cytoplasmic adapter protein; Lymphocyte antigen 57; MGC111051; SLP-65; Src homology 2 domain-containing leukocyte protein of 65 kDa; Src homology [SH2] domain-containing leukocyte protein of 65 kD
Gene Aliases: AGM4; BASH; Bca; BLNK; BLNK-S; Ly-57; LY57; Lyw-57; SLP-65; SLP65
UniProt ID: (Human) Q8WV28, (Mouse) Q9QUN3
Entrez Gene ID: (Human) 29760, (Mouse) 17060
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support