Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
The antibody is specific to NLOC1S1.
BLOC1S1 is a component of the ubiquitously expressed BLOC1 multisubunit protein complex. BLOC1 is required for normal biogenesis of specialized organelles of the endosomal-lysosomal system, such as melanosomes and platelet dense granules (Starcevic and Dell'Angelica, 2004 ).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: biogenesis of lysosomal organelles complex-1, subunit 1; Biogenesis of lysosome-related organelles complex 1 subunit 1; biogenesis of lysosome-related organelles complex-1, subunit 1; BLOC subunit 1; BLOC-1 subunit 1; GCN5 (general control of amino-acid synthesis, yeast, homolog)-like 1; GCN5 general control of amino acid synthesis-like 1; GCN5 general control of amino-acid synthesis 5-like 1; GCN5-like protein 1; general control of amino acid synthesis, yeast homolog-like 1; general control of amino acid synthesis-like 1; MTA1-interacting coactivator; Protein RT14
Gene Aliases: AI839753; BLOC1S1; BLOS1; BORCS1; GCN5L1; MICoA; RT14
UniProt ID: (Human) P78537, (Mouse) O55102, (Rat) D3ZKU7
Entrez Gene ID: (Human) 2647, (Mouse) 14533, (Rat) 288785
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support