Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a protein that is capable of inducing formation of cartilage in vivo. Although other bone morphogenetic proteins are members of the TGF-beta superfamily, this gene encodes a protein that is not closely related to other known growth factors. This gene is expressed as alternatively spliced variants that share an N-terminal protease domain but differ in their C-terminal region.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BMP; BMP-1; Bone morphogenetic protein; Bone morphogenetic protein 1; FLJ44432; Mammalian tolloid protein; mTld; PCP; pCP-2; procollagen C-endopeptidase; Procollagen C-proteinase; procollagen C-proteinase 3
Gene Aliases: BMP1; OI13; PCOLC; PCP; PCP2; TLD
UniProt ID: (Human) P13497
Entrez Gene ID: (Human) 649
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support