Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a secreted protein that interacts with, and inhibits bone morphogenetic protein (BMP) function. It has been shown to inhibit BMP2- and BMP4-dependent osteoblast differentiation and BMP-dependent differentiation of the chondrogenic cells. Mutations in this gene are associated with a lethal skeletal disorder, diaphanospondylodysostosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BMP-binding endothelial regulator precursor protein; BMP-Binding Endothelial Regulator Protein; Bone morphogenetic protein-binding endothelial cell precursor-derived regulator; CRIM3; crossveinless 2; crossveinless-2; hCV2; mCV2; Protein crossveinless-2
Gene Aliases: 3110056H04Rik; BMPER; CRIM3; CV-2; CV2; KIAA1965; RGD1563373
UniProt ID: (Human) Q8N8U9, (Mouse) Q8CJ69
Entrez Gene ID: (Human) 168667, (Mouse) 73230, (Rat) 300455
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support