Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Recommended positive controls: A549, HeLa, HepG2, HCT116, mouse liver.
Predicted reactivity: Mouse (86%), Rat (87%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Component of the PeBoW complex, which is required for maturation of 28S and 5.8S ribosomal RNAs and formation of the 60S ribosome.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Block of proliferation 1 protein; Ribosome biogenesis protein BOP1
Gene Aliases: AU020183; AW146150; BOP1; D18861; Erb1p; KIAA0124; mKIAA0124
UniProt ID: (Human) Q14137, (Mouse) P97452
Entrez Gene ID: (Human) 23246, (Mouse) 12181
Molecular Function:
ribosomal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support