Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene is located in the nonrecombining portion of the Y chromosome, and expressed specifically in testis. The encoded protein interacts with ubiquitin protein ligase E3A and may be involved in male germ cell development and male infertility. Three nearly identical copies of this gene exist on chromosome Y; two copies are part of a palindromic region. This record represents the copy outside of the palidromic region.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Basic charge, Y-linked 2; basic protein on Y chromosome 2; testis-specific basic protein on Y, 2; Testis-specific basic protein Y 2; variable charge, Y-linked, 2; Variably charged protein Y 2; y-linked
Gene Aliases: BPY2; BPY2A; BPY2B; BPY2C; VCY2; VCY2A; VCY2B; VCY2C
UniProt ID: (Human) O14599
Entrez Gene ID: (Human) 9083, (Human) 442868
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support