Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
Required for correct progression through G2 phase of the cell cycle and entry into mitosis. Required for RCOR1/CoREST mediated repression of neuronal specific gene promoters.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 94 kDa B-raf protein; B-Raf proto-oncogene serine/threonine-protein kinase (p94); BRCA2-associated factor 35; FLJ26127; high-mobility group 20B; HMG box domain containing 2; HMG box-containing protein 20B; HMG domain-containing protein 2; HMG domain-containing protein HMGX2; murine sarcoma viral (v-raf) oncogene homolog B1; p94; Proto-oncogene B-Raf; Serine/threonine-protein kinase B-raf; SMARCE1-related protein; Sox-like transcriptional factor; Structural DNA-binding protein BRAF35; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily E member 1-related; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily E, member 1-related; v-raf murine sarcoma viral oncogene homolog B; v-Raf murine sarcoma viral oncogene homolog B1
Gene Aliases: B-raf; B-RAF1; BRAF; BRAF1; BRAF25; BRAF35; HMG20B; HMGX2; HMGXB2; NS7; PP7706; pp8857; RAFB1; SMARCE1R; SOXL
UniProt ID: (Human) P15056, (Human) Q9P0W2
Entrez Gene ID: (Human) 673, (Human) 10362
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support