Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Primary Antibodies
  • Search All
  • Popular Product Areas
  • TaqMan Assays
  • Primary Antibodies
  • Secondary Antibodies
  • Isotype Controls
  • Proteins and Peptides
  • ELISA Kits
  • All Documents & Support
  • Certificates
  • SDS
  • Product FAQs
  • Documents & Support
  • Manuals & Protocols
    Search button Close
            Target of Interest
            Application
            Target Species
            • Order Status
            • 快速訂購
            • 登入
              登入
              Don't have an account ? Create Account
              • 帳戶
              • 訂單
              • 訂製產品與專案

            Disclaimer

            Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

            Loading ...
            • Primary Antibodies ›
            • BRD4 Antibodies

            Invitrogen

            BRD4 Recombinant Rabbit Monoclonal Antibody (14H4L4)

            1 Reference
            View all (28) BRD4 antibodies
            Datasheet
            Protocols
            Questions & Answers
            Tech Support
            Promotion Available
            Learn more
            Catalog # 702448
            Web quote
            You can generate a web quote (PDF) by signing in to your account and adding products to your cart. View cart and click the 'Generate a web quote' button. With your web quote document, you can get approval for your order and complete your order online.
             
            Price (HKD)
            4,133.00
            100 µg
             
            Check your price & availability
            Request bulk or custom format
            Datasheet
            Protocols
            Questions & Answers
            Tech Support

            Cite BRD4 Recombinant Rabbit Monoclonal Antibody (14H4L4)

            • Antibody Testing Data (3)
            Application
            BRD4 Antibody in Immunocytochemistry (ICC/IF)
            Group 53 Created with Sketch.

            FIGURE: 1 / 3

            BRD4 Antibody (702448) in ICC/IF

            For immunofluorescence analysis, HeLa cells were fixed and permeabilized for detection of endogenous BRD4 using Anti- BRD4 Recombinant Rabbit Monoclonal Antibody (Product # 702448, 2 µg/mL) and labeled with Goat anti-Rabbit IgG (Heavy Chain) Superclonal™ Secondary Antibody, Alexa Fluor® 488 conjugate (Product # A27034, 1:2000). Panel a) shows representative cells that were stained for detection and localization of BRD4 protein (green), Panel b) is stained for nuclei (blue) using SlowFade® Gold Antifade Mountant with DAPI (Product # S36938)... View More
            View Product

            BRD4 Antibody in Immunocytochemistry (ICC/IF)
            BRD4 Antibody in Western Blot (WB)
            BRD4 Antibody in ChIP Assay (ChIP)

            Product Details

            702448

            Applications
            Tested Dilution
            Publications

            Western Blot (WB)

            1-2 µg/mL
            View 1 publication 1 publication

            Immunocytochemistry (ICC/IF)

            2 µg/mL
            -

            ChIP assay (ChIP)

            2.5 µg/1x10^6 cells
            -
            Product Specifications

            Species Reactivity

            Human

            Published species

            Human

            Host/Isotype

            Rabbit / IgG

            Expression System

            Expi293

            Class

            Recombinant Monoclonal

            Type

            Antibody

            Clone

            14H4L4

            Immunogen

            Protein corresponding to human BRD4 (aa154-aa284)
            View immunogen

            Conjugate

            Unconjugated

            Form

            Liquid

            Concentration

            0.5 mg/mL

            Purification

            Protein A

            Storage buffer

            PBS, pH 7.4

            Contains

            0.09% sodium azide

            Storage conditions

            Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

            Shipping conditions

            Wet ice

            RRID

            AB_2688043

            Product Specific Information

            This antibody is predicted to react with Monkey, Rat, Horse, Bovine

            Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

            Target Information

            BRD4, a ubiquitously expressed nuclear phosphoprotein, is a member of BET protein family. It is a human homolog of mouse MCAP and consists of 2 N-terminal bromodomains, a central ET-domain, and several kinase-like motifs in its N-terminus. It is expressed as 2 isoforms; the longer variant has a C-terminus region with a praline and glutamine-rich domains. BRD4 is a chromatin-binding protein whose expression is induced in response to growth stimuli. BRD4 acts at different stages of the cell cycle by interacting with proteins including RFC and SPA-1 and regulates cell growth. It also has a role in regulating chromosomal dynamics during mitosis. BRD4 functions as a cellular target for E2 protein of papillomavirus.

            For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

            Bioinformatics

            Protein Aliases: BRD4-NUT FUSION; bromodomain-containing 4; Bromodomain-containing protein 4; chromosome-associated protein; Protein HUNK1

            View more View less

            Gene Aliases: BRD4; CAP; HUNK1; HUNKI; MCAP

            View more View less

            UniProt ID: (Human) O60885

            View more View less

            Entrez Gene ID: (Human) 23476

            View more View less

            Function(s)

            p53 binding chromatin binding protein binding lysine-acetylated histone binding

            Process(es)

            regulation of transcription involved in G1/S transition of mitotic cell cycle transcription, DNA-templated cellular response to DNA damage stimulus positive regulation of G2/M transition of mitotic cell cycle viral process covalent chromatin modification positive regulation of transcription elongation from RNA polymerase II promoter positive regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of transcription from RNA polymerase II promoter regulation of inflammatory response regulation of phosphorylation of RNA polymerase II C-terminal domain
            It has to be done as per old AB suggested Products section.
            guarantee_icon

            Performance Guarantee

            If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

            Learn more
            help_icon

            We're here to help

            Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

            Contact tech support
            Ordering Plus Icon Minus Icon
            • Order Status
            • Order Help
            • Quick Order
            • Supply Center
            • eProcurement
            Support Plus Icon Minus Icon
            • Help and Support
            • Contact Us
            • Technical Support Centers
            • Documents and Certificates
            • Report a Site Issue
            Resources Plus Icon Minus Icon
            • Learning Centers
            • Promotions
            • Events and Webinars
            • Social Media
            About Thermo Fisher Plus Icon Minus Icon
            • About Us About Us
            • Careers Careers
            • Investors Investors
            • News News
            • Social Responsibility Social Responsibility
            • Trademarks
            Our Portfolio Plus Icon Minus Icon
            • Thermo Scientific
            • Applied Biosystems
            • Invitrogen
            • Gibco
            • Ion Torrent
            • Fisher Scientific
            • Unity Lab Services
            • Patheon
            • PPD
            • Terms & Conditions
            • Privacy Policy
            • Price & Freight Policy
            • Cookie偏好

              在此網站使用 Cookie 的選擇

              我們以及我們的附屬公司和供應商使用 cookies 和類似技術來經營我們的網站、識別我們網站的訪客、提供安全登入、收集統計資料以優化網站功能,並提供根據您的興趣量身定制的內容。點擊「全部接受」 以接受所有 cookies,並直接進入網站,或點擊「管理設定」 以查看 cookies 類型的詳細說明,並選擇是否在網站接受某些 cookies。
            © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
            hong kong flag icon
            Hong Kong

            Your items have has been added!


            Host server : magellan-search-green-7c4bf66445-pkshg:80/100.66.78.18:80.
            git-commit: 01fb56a08528fd72f4925b2983aa4df42f9d7994
            git-url: https://github.com/thermofisher/magellan-search
            git-branch: release/2.30.1-2025.05.22-1.0