Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
RNA polymerase (pol) III synthesizes tRNA, 5s rRNA, 7SL RNA and U6 snRNA and is overexpressed in many transformed cell lines and tumors in vivo, since cells must duplicate its protein components before division. Therefore, in order to maintain rapid growth, cells must produce a high level of Pol III transcribed RNA, which requires the presence of the TFIIIB and TFIIIC2 transcription factor complexes. The TFIIIC2 complex is composed of five subunits, TFIIIC220, TFIIIC110, TFIIIC102, TFIIIC90 and TFIIIC63, that are overexpressed in adenovirus transformed cells as well as in malignant cells in vivo, such as ovarian carcinomas. TFIIIC2 recruits RNA pol III and TFIIIB to promoter elements and may be a key component in the deregulation of malignant cells. The TFIIIB complex includes the TATA-binding protein (TBP), TFIIB-related factor 1 (TFIIIB90, BRF1) and TFIIIB, the expression of which are also upregulated in transformed cells. In many carcinomas, the tumor suppressors retinoblastoma (RB) and p53 are inactivated, which affects their ability to bind and inactivate the function of TFIIIB.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B - related factor 1; B-related factor 1; BRF-1; BRF1 homolog, subunit of RNA polymerase III transcription initiation factor IIIB; epididymis secretory sperm binding protein Li 76p; general transcription factor IIIB, 90kD subunit; hTFIIIB90; TAF3B2; TATA box binding protein (TBP)-associated factor 3C; TATA box binding protein (TBP)-associated factor, RNA polymerase III, GTF3B subunit 2; TATA box-binding protein-associated factor, RNA polymerase III, subunit 2; TBP - associated factor, RNA polymerase III, 90kD; TFIIIB90; Transcription factor IIIB 90 kDa subunit
Gene Aliases: 2510002F24Rik; BRF; BRF-1; BRF1; CFDS; GTF3B; hBRF; HEL-S-76p; mTFIIIB90; TAF3B2; TAF3C; TAFIII90; TF3B90; TFIIIB90
UniProt ID: (Human) Q92994
Entrez Gene ID: (Human) 2972, (Mouse) 72308
Molecular Function:
general transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support