Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes one of the multiple subunits of the RNA polymerase III transcription factor complex required for transcription of genes with promoter elements upstream of the initiation site. The product of this gene, a TFIIB-like factor, is directly recruited to the TATA-box of polymerase III small nuclear RNA gene promoters through its interaction with the TATA-binding protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B-related factor 2; BRF-2; BRF2, subunit of RNA polymerase III transcription initiation factor, BRF1-like; FLJ11052; hBRFU; hTFIIIB50; RNA polymerase III transcription initiation factor BRFU; TFIIIB50; transcription factor IIB- related factor, TFIIIB50; Transcription factor IIIB 50 kDa subunit
Gene Aliases: 2700059M06Rik; 5730512K07Rik; AU018504; AU022602; BRF2; BRFU; PRO1470; TFIIIB50
UniProt ID: (Human) B4DFZ6, (Rat) Q4V8D6, (Mouse) Q3UAW9
Entrez Gene ID: (Human) 55290, (Rat) 306542, (Mouse) 66653
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support