Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MRKPRRKSRQ NAEGRRSPSP YSLKCSPTRE TLTYAQAQRI VEVDIDGRLH RISIYDPLKI ITEDELTAQD ITECNSNKEN SEQPQFPGKS KKPSSKGKKK ESCSKHASGT SFHLPQPSFR MVDSGIQPEA PPLPAAYYRY IEKPPEDLDA EVEYDMDEED LAWLDMVNEK RRVDGHSLVS ADTFELLVDR LEKESYLESR SSGAQQSLID EDAFCCVCLD DECHNSNVIL FCDICNLAVH QECYGVPYIP EGQWLCRCCL QSPSRPVDCI LCPNKGGAFK QTSDGHWAHV VCAIWIPEVC FANTVFLEPI EGIDNIPPAR WKLTCYICKQ KGLGAAIQCH KVNCYTAFHV TCAQRAGLFM KIEPMRETSL NGTIFTVRKT AYCEAHSPPG AATARRKGDS PRSISETGDE EGLKEGDGEE EEEEEVEEEE QEAQGGVSGS LKGVPKKSKM SLKQKIKKEP EEAGQDTPST LPMLAVPQIP SYRLNKICSG LSFQRKNQFM QRLHNYWLLK RQARNGVPLI RRLHSHLQSQ RNAEQREQDE KTSAVKEELK YWQKLRHDLE RARLLIELIR KREKLKREQV KVQQAAMELE LMPFNVLLRT TLDLLQEKDP AHIFAEPVNL SEVPDYLEFI SKPMDFSTMR RKLESHLYRT LEEFEEDFNL IVTNCMKYNA KDTIFHRAAV RLRDLGGAIL RHARRQAENI GYDPERGTHL PESPKLEDFY RFSWEDVDNI LIPENRAHLS PEVQLKELLE KLDLVSAMRS SGARTRRVRL LRREINALRQ KLAQPPPPQP PSLNKTVSNG ELPAGPQGDA AVLEQALQEE PEDDGDRDDS KLPPPPTLEP TGPAPSLSEQ ESPPEPPTLK PINDSKPPSR FLKPRKVEED ELLEKSPLQL GNEPLQRLLS DNGINRLSLM APDTPAGTPL SGVGRRTSVL FKKAKNGVKL QRSPDRVLEN GEDHGVAGSP ASPASIEEER HSRKRPRSRS CSESEGERSP QQEEETGMTN GFGKHTESGS DSECSLGLSG GLAFEACSGL TPPKRSRGKP ALSRVPFLEG VNGDSDYNGS GRSLLLPFED RGDLEPLELV WAKCRGYPSY PALIIDPKMP REGLLHNGVP IPVPPLDVLK LGEQKQAEAG EKLFLVLFFD NKRTWQWLPR DKVLPLGVED TVDKLKMLEG RKTSIRKSVQ VAYDRAMIHL SRVRGPHSFV TSSYL
Scaffold subunit of various histone acetyltransferase (HAT) complexes, such as the MOZ/MORF and HBO1 complexes, which have a histone H3 acetyltransferase activity (PubMed:16387653, PubMed:26620551, PubMed:26677226). Plays a role in DNA replication initiation by directing KAT7/HBO1 specificity towards histone H3 'Lys-14' acetylation (H3K14ac), thereby facilitating the activation of replication origins (PubMed:26620551). Component of the MOZ/MORF complex which has a histone H3 acetyltransferase activity (PubMed:16387653). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bromodomain and PHD finger containing, 3; Bromodomain and PHD finger-containing protein 3
Gene Aliases: BRPF3; KIAA1286
UniProt ID: (Human) Q9ULD4
Entrez Gene ID: (Human) 27154
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support