Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute at 0.2 mg/mL in sterile PBS.
This gene encodes a member of the trypsin family of serine proteases. The enzyme is expressed in the airways in a developmentally regulated manner. The gene is part of a cluster of serine protease genes on chromosome 16.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Brain-specific serine protease 4; BSP-2; BSSP-4; MGC9599; protease, serine, 22; Serine protease 22; Serine protease 26; Tryptase epsilon
Gene Aliases: 4733401N09Rik; BSSP-4; Bssp4; Prss22; Prss26; SP001LA
UniProt ID: (Mouse) Q9ER10
Entrez Gene ID: (Mouse) 70835
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support