Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
FIGURE: 1 / 2
In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse Tryptase epsilon /BSSP-4 is observed.
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
This gene encodes a member of the trypsin family of serine proteases. The enzyme is expressed in the airways in a developmentally regulated manner. The gene is part of a cluster of serine protease genes on chromosome 16.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Brain-specific serine protease 4; BSP-2; BSSP-4; MGC9599; prosemin; protease, serine S1 family member 22; protease, serine, 22; Serine protease 22; Serine protease 26; Tryptase epsilon
Gene Aliases: BSSP-4; BSSP4; hBSSP-4; PRSS22; PRSS26; SP001LA; UNQ302/PRO343
UniProt ID: (Human) Q9GZN4
Entrez Gene ID: (Human) 64063
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support