Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen Sequence: MESAAALHFS RPASLLLLLL SLCALVSAQF IVVGPTDPIL ATVGENTTLR CHLSPEKNAE DMEVRWFRSQ FSPAVFVYKG GRERTEEQME EYRGRTTFVS
The specific functin of this protein remains unknown. This gene is a member of the BTN2 subfamily of genes, which encode proteins belonging to the butyrophilin protein family. The gene is located in a cluster on chromosome 6, consisting of seven genes belonging to the expanding B7/butyrophilin-like group, a subset of the immunoglobulin gene superfamily. The encoded protein is an integral plasma membrane B box protein involved in lipid, fatty-acid and sterol metabolism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Butyrophilin; Butyrophilin subfamily 2 member A1; butyrophilin, subfamily 2, member A1; FLJ36567
Gene Aliases: BK14H9.1; BT2.1; BTF1; BTN2.1; BTN2A1; DJ3E1.1
UniProt ID: (Human) Q7KYR7
Entrez Gene ID: (Human) 11120
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support