Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
BTN3A2 encodes a member of the immunoglobulin superfamily, which resides in the juxta-telomeric region of the major histocompatability class 1 locus and is clustered with the other family members on chromosome 6. The encoded protein may be involved in the adaptive immune response. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: butyrophilin protein; Butyrophilin subfamily 3 member A2; butyrophilin, subfamily 3, member A2
Gene Aliases: BT3.2; BTF3; BTF4; BTN3.2; BTN3A2; CD277
UniProt ID: (Human) P78410
Entrez Gene ID: (Human) 11118
Molecular Function:
immunoglobulin receptor superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support