Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDTLDRVVKP KTKRAKRFLE KREPKLNENI KNAMLIKGGN ANATVTKVLK DVYALKKPYG VLYKKKNITR PFEDQTSLEF FSKKSDCSLF MFGSHNKKRP NNLVIGRMYD YHVLDMIELG IENFVSLKDI KNSKCPEGTK PMLIFAGDDF DVTEDYRRLK SLLIDFFRGP TVSNIRLAGL EYVLHFTALN GKIYFRSYKL LLKKSGCRTP RIELEEMGPS LDLVLRRTHL ASDDLYKLSM KMPKALKPKK KKNISHDTFG TTYGRIHMQK QDLSKLQTRK MKGLKKRPAE RITEDHEKKS KRIKKN
RPF2 is a protein coding gene involved in ribosomal large subunit assembly. May regulate the localization of the 5S RNP/5S ribonucleoprotein particle to the nucleolus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: brix domain containing 1; Brix domain-containing protein 1; homolog of Rpf2; ribosomal processing factor 2 homolog; Ribosome biogenesis protein RPF2 homolog; Ribosome production factor 2 homolog; RP11-397G5.1
Gene Aliases: bA397G5.4; BXDC1; RPF2
UniProt ID: (Human) Q9H7B2
Entrez Gene ID: (Human) 84154
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support