Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
FIGURE: 1 / 3
The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
Based on 100% sequence identity, this antibody is predicted to react with Mouse
This gene encodes a C2H2-type zinc finger protein and is closely related to BCL11A, a gene whose translocation may be associated with B-cell malignancies. The specific function of this gene has not yet been determined. Two alternatively spliced transcript variants, which encode distinct isoforms, have been reported.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B-cell CLL/lymphoma 11B; B-cell CLL/lymphoma 11B (zinc finger protein); B-cell CLL/lymphoma 11B/T-cell receptor delta constant region fusion protein; B-cell lymphoma/leukaemia 11B; B-cell lymphoma/leukemia 11B; BCL-11B; BCL11B/TRDC fusion; COUP-TF-interacting protein 2; hRit1; Radiation-induced tumor suppressor gene 1 protein; zinc f; zinc finger protein hRit1 alpha
Gene Aliases: ATL1; ATL1-alpha; ATL1-beta; ATL1-delta; ATL1-gamma; BCL11B; CTIP-2; CTIP2; hRIT1-alpha; RIT1; ZNF856B
UniProt ID: (Human) Q9C0K0
Entrez Gene ID: (Human) 64919
Molecular Function:
DNA-binding transcription factor
zinc finger transcription factor
C2H2 zinc finger transcription factor
gene-specific transcriptional regulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support