Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: CI2Z9E .
Suggested positive control: A431 and HEK kidney cell lysates and MCC827 lung cell lysates, A-431 whole cell lysate1, 293 whole cell lysate.
Involved in 2 distinct lysosomal degradation pathways: acts as a regulator of autophagy and as a regulator of G-protein coupled receptors turnover. Regulates degradation in lysosomes of a variety of G-protein coupled receptors via its interaction with GPRASP1/GASP1. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Bcl-2; BCS2; beclin 1, autophagy related, pseudogene 1; Beclin-1 autophagy-related pseudogene 1; Beclin-1-like protein 1; Beclin-2; PFM 2; PFM2; PRDM 5; PRDM5; PRDM5 protein
Gene Aliases: BECN1L1; BECN1P1; BECN2
UniProt ID: (Human) A8MW95
Entrez Gene ID: (Human) 441925
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support