Search Thermo Fisher Scientific
Rockland
Promotions
View available promotion(s)
Promo Code: P5877545 Don't miss out: buy 3, only pay for 2 From June 2-13 take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Store vial at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.
This antiserum detects the minimal peptide substrate (GLNDIFEAQKIEWH) for the E. coli biotin ligase tag (BLT of BirA). Western blot analysis of B. subtilis strains producing either (BAP-tagged) SPoIIIAH (XWB49) or (BAP-tagged) SpoIIQ (XWB46) showed strong and specific reactivity to the antisera. Minor background bands of unknown identity may be observed. The antisera will recognize the BAP/BLT tag irrespective of the whether the tag is biotinylated (data not shown).
The peptide is also called the biotin acceptor peptide (BAP) of the biotin carboxyl carrier protein (BCCP) subunit of acetyl CoA carboxylase of E. coli.