Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: RRSYNVTPPP IEESHPYYQE IYNDRRYKVC DVPLDQLPRS ESLKDVLERL LPYWNERIAP EVLRGKTILI SAHGNSSRAL LKHLEGISDE DIINITLPTG VPILLELDEN LRAVGPHQFL GDQEAIQAAI KKVEDQGKVK QA
Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 90%.
Riboflavin kinase is an essential enzyme that catalyzes the phosphorylation of riboflavin to form flavin mononucleotide, an obligatory step in vitamin B2 utilization and flavin cofactor synthesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2,3-bisphosphoglycerate mutase; 2,3-bisphosphoglycerate mutase, erythrocyte; 2,3-bisphosphoglycerate synthase; 2,3-diphosphoglycerate mutase; Bisphosphoglycerate mutase; BPG-dependent PGAM; BPGM; DPGM; erythrocyte 2,3-bisphosphoglycerate mutase; testis secretory sperm-binding protein Li 202a
Gene Aliases: Ab2-098; AI323730; AL022789; BPGM; C86192; DPGM
UniProt ID: (Human) P07738, (Mouse) P15327
Entrez Gene ID: (Human) 669, (Rat) 296973, (Mouse) 12183
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support