Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Endotoxin level < 0.01EU/µg by LAL method
Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Briakinumab is a human anti-IL-12 monoclonal antibody being developed for the treatment of a number of T-cell driven autoimmune diseases. It targets and neutralize interleukin-12 and interleukin-23, two proteins associated with inflammation, such as pro-inflammatory interleukins or tumor necrosis factor- alpha. Briakinumab represents a novel approach to treating psoriasis, Multiple Sclerosis, Crohn's Disease and other autoimmune and inflammatory disorders. As of 2011 pharmaceutical development for psoriasis has been discontinued in the U. S. and Europe. Briakinumab targets and neutralizes interleukin-12 and interleukin-23.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ABT-874
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support