Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive Control: A549, HL-60, A431, rat colon tissue lysate, LOVO, rat testis tissue, human thyroid gland tissue, human colon cancer tissue, mouse fallopian tissue, PC-3M.
This gene encodes a protein involved in spindle checkpoint function. The encoded protein contains four WD repeat domains and has sequence similarity with the yeast BUB3 protein. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BUB3 budding uninhibited by benzimidazoles 3 homolog; BUB3 mitotic checkpoint protein; budding uninhibited by benomyl; budding uninhibited by benzimidazoles 3 homolog; mitotic checkpoint component; Mitotic checkpoint protein BUB3; OTTHUMP00000020682; testicular tissue protein Li 27
Gene Aliases: BUB3; BUB3L; hBUB3
UniProt ID: (Human) O43684
Entrez Gene ID: (Human) 9184
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support