Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QSPWCSQYR RQLLKSLPEL SASAELCIED RPMPKLEIEK EIELGNEDYC IKREYLICED YKLFWVAPRN SAELTVIKVS SQPVPWDFYI NLKLKERLNE DFDHFCSCYQ YQDGCIVWHQ YINCFTLQDL LQHSEYITHE ITVLIIYNLL TIVEMLHKAE IVHGDLSPRC LILRNRIHDP YDCNKNNQAL KIVDFSYSVD LRVQLDVFTL SGFRTVQILE GQKILANCSS PYQVDLFGIA DLAHLLLFKE HLQVFWDGSF WKLSQNISEL KDGELWNKFF VRILNANDEA TVSVLGELAA EMNGVFDTTF QSHLNKALWK VGKLTSPGAL LFQ (719-1050 aa encoded by B C018739)
BUBR1 is a member of the Ser/Thr protein kinase family. It is a component of the mitotic checkpoint that delays anaphase until all chromosome are properly attached to the mitotic spindle. Compromised spindle checkpoints are thought to play an important role in genetic instability that predisposes cells to malignant transformations. Human cells contain two related checkpoint protein kinases, evolved from the BUB1 gene; hBUBR1 and hBUB1. They are important components of the spindle checkpoint Both kinases monitor mitotic kinetochore-microtubule interactions. hBUBR1 is essential for normal mitotic progression as it prevents cells from entering into anaphase too early. BUBR1 has been detected in human cancers. It has been identified as a novel binding partner of BCSG1 (Breast Cancer-Specific Gene 1), which has been suggested to accelerate the progression of breast cancer by compromising the mitotic checkpoint control through inactivation of BUBR1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BUB1B, mitotic checkpoint serine/threonine kinase; BubR1; budding uninhibited by benzimidazoles 1 beta; budding uninhibited by benzimidazoles 1 homolog beta; hBub1; hBUBR1; MAD3/BUB1-related protein kinase; Mitotic checkpoint kinase MAD3L; Mitotic checkpoint serine/threonine-protein kinase BUB1 beta; Protein SSK1
Gene Aliases: AU045529; Bub1A; BUB1B; BUB1beta; BUBR1; hBUBR1; MAD3L; MVA1; SSK1
UniProt ID: (Human) O60566, (Mouse) Q9Z1S0
Entrez Gene ID: (Human) 701, (Mouse) 12236
Molecular Function:
non-receptor serine/threonine protein kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support