Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
MA1-825 detects C/EBP alpha protein in human, mouse and rat samples.
MA1-825 has successfully been used in Western blot and immunoprecipitation procedures. By Western blot, this antibody detects a 45 kDa protein representing C/EBP alpha in transfected hu293T cells. MA1-825 is specific to the amino terminal region of C/EBP alpha.
The MA1-825 immunogen is a synthetic peptide corresponding to residues M(1) E S A D F Y E V E P R P P(14) of mouse C/EBP alpha. This peptide (Cat. # PEP-242) is available for use in neutralization and control experiments.
The C/EBP (CCAAT/enhancer binding protein) family of transcription factors--alpha, beta, delta, and epsilon)--are widely expressed proteins found in a variety of tissues. These domains contain an evolutionarily conserved five-amino acid motif (the regulatory domain motif RDM) that conforms to the consensus sequence (I/V/L) KXEP. One of the major roles of these transcription factors is that of metabolic regulation in liver and fat cells. Studies suggest that these transcription factors act in a cascade aiding in the development of the adipose cell. Three C/EBP beta isoforms (LAP*, LAP, and LIP) arise from different translation initiation and display different function in gene regulation. From a neurological standpoint, C/EBPs have been implicated in the mediation of nerve growth factor, altering synaptic plasticity, and mediation of inflammatory response in AD patients.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C EBP; C/EBP alpha; CAAT/enhancer-binding protein DNA-binding protein; CAAT/enhancer-binding protein, DNA-binding protein; CCAAT/enhancer binding protein (C/EBP), alpha; CCAAT/enhancer binding protein, alpha; CCAAT/enhancer-binding protein alpha; OTTHUMP00000220269
Gene Aliases: C/EBP-alpha; C/ebpalpha; CBF-A; CEBP; CEBPA; DBPCEP
UniProt ID: (Human) P49715, (Mouse) P53566, (Rat) P05554
Entrez Gene ID: (Human) 1050, (Mouse) 12606, (Rat) 24252
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support