Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 1
Positive Samples: Human plasma, Mouse plasma, Rat plasma
Immunogen sequence: SLKLYHAFSA MKKVETNMAF SPFSIASLLT QVLLGAGENT KTNLESILSY PKDFTCVHQA LKGFTTKGVT SVSQIFHSPD LAIRDTFVNA SRTLYSSSPR V
C1-inhibitor (C1INH) functions as a regulator of the activation of the classical complement pathway and of the contact activation system of kinin generation and coagulation. Its primary biologically relevant target proteinases are C1r, C1s, coagulation factors XIa and XIIa, and plasma kallikrein. C1-inhibitor consists of one polypeptide chain and has a molecular mass of ~104 kDa. Serum concentration has been determined to 180 µg/mL.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C1 esterase inhibitor; C1 Inh; C1 inhibitor; C1-inhibiting factor; C1Inh; complement component 1 inhibitor; esterase inhibitor; IC1; Plasma protease C1 inhibitor; serine (or cysteine) peptidase inhibitor, clade G, member 1; serine (or cysteine) proteinase inhibitor, clade G (C1 inhibitor), member 1; serine (or cysteine) proteinase inhibitor, clade G (C1 inhibitor), member 1, (angioedema, hereditary); serine (or cysteine) proteinase inhibitor, clade G, member 1; serine/cysteine proteinase inhibitor clade G member 1; Serpin G1; serpin peptidase inhibitor clade G member 1; serpin peptidase inhibitor, clade G (C1 inhibitor), member 1; serpin peptidase inhibitor, clade G, member 1
Gene Aliases: C1IN; C1INH; C1NH; HAE1; HAE2; SERPING1
UniProt ID: (Human) P05155, (Rat) Q6P734, (Mouse) P97290
Entrez Gene ID: (Human) 710, (Rat) 295703, (Mouse) 12258
Molecular Function: protease inhibitor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support