Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EELDQYKDFGF HKMESFAKET SSPKSNLEKE NLEYKVENER LLQIISELEE EIQINLKENS GLEDELISMK EMAEKDHKTI QK
Highest antigen sequence identity to the following orthologs - mouse 55%, rat 52%.
C10orf67 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: long intergenic non-protein coding RNA 1552; uncharacterized protein C10orf67; Uncharacterized protein C10orf67, mitochondrial
Gene Aliases: bA215C7.4; C10orf115; C10orf67; LINC01552
UniProt ID: (Human) Q8IYJ2
Entrez Gene ID: (Human) 256815
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support