Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PGLQESHLKE TPSLRRVHLG TGACPWSGSF PLENTELANV GANQVTVRKG EKDHTTHCHA SDHANQLSIH IPGWSYRAVH TKVFSGSSKR QQGEVCM
Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 48%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: (E2-independent) E3 ubiquitin-conjugating enzyme FATS; Centrosomal protein C10orf90; E2/E3 hybrid ubiquitin-protein ligase FATS; FATS; Fragile-site associated tumor suppressor homolog
Gene Aliases: bA422P15.2; C10orf90; FATS
UniProt ID: (Human) Q96M02
Entrez Gene ID: (Human) 118611
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support