Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cilia and flagella associated protein 46; Cilia- and flagella-associated protein 46; protein CFAP46; tetratricopeptide repeat domain 40; Tetratricopeptide repeat protein 40; TPR repeat-containing protein C10orf93
Gene Aliases: 9330101J02Rik; bA288G11.4; bA288G11.5; bB137A17.2; bB137A17.3; C10orf123; C10orf124; C10orf92; C10orf93; CFAP46; E030019B06Rik; Gm170; RGD1559985; TTC40
UniProt ID: (Human) B7WPK8
Entrez Gene ID: (Human) 54777, (Mouse) 212124, (Rat) 499282
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support