Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RSGLWVFCY RDRLVMQKGL KDNFADVQVS VVDCPDLTKE PFTFPVKGIC GKTRIAEVGG VPYLLPLVNQ KKVYDLNKIA KEIKLPGAFI LGAGAGPFQT LGFNSEFMPV IQTESEHKPP VNGSYFAHVN PADGGCLLEK YSEKCHDFQC ALLANLFASE GQPGKVIEVK AKRRTGPLNF VTCMRETLEK HYGNKPIGMG GTFIIQKGKV KSHIMPAEFS SCPLNSDEEV NKWLHFYEMK APLVCLPVFV SRDPGFDLRL EHTHFFSRHG EGGHYHYDTT PDIVEYLGYF LPAEFLYRID QPKETHSIGR D (1-310 aa encoded by BC007110)
C11orf54 protein function is currently unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ester hydrolase C11orf54; Ester hydrolase C11orf54 homolog
Gene Aliases: C11orf54; LP4947; PTD012; PTOD012
UniProt ID: (Human) Q9H0W9, (Rat) Q5U2Q3
Entrez Gene ID: (Human) 28970, (Rat) 363016
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support