Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: CPDLTKEPFT FPVKGICGKT RIAEVGGVPY LLPLVNQKKV YDLNKIAKEI
Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 92%; Human: 100%; Mouse: 100%; Rabbit: 86%; Rat: 100%; Zebrafish: 79%
C11orf54 protein function is currently unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ester hydrolase C11orf54
Gene Aliases: C11orf54; LP4947; PTD012; PTOD012
UniProt ID: (Human) Q9H0W9
Entrez Gene ID: (Human) 28970
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support