Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSAARESHPH GVKRSASPDD DLGSSNWEAA DLGNEERKQK FLRLMGAGKK EHTGRLVIGD HKSTSHFRTG EEDKKINEEL ESQYQQSMDS KLSGRYRRHC GLGFSEVEDH DGEGDVAGDD DDDDDDSPDP ESPDDSESDS ESEKEESAEE LQAAEHPDEV EDPKNKKDAK SNYKMMFVKS SGS
C11orf58 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Small acidic protein
Gene Aliases: C11orf58; IMAGE145052; SMAP
UniProt ID: (Human) O00193
Entrez Gene ID: (Human) 10944
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support