Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PPP1R32 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein phosphatase 1 regulatory subunit 32; protein phosphatase 1, regulatory subunit 32; Stabilizer of axonemal microtubules 4
Gene Aliases: C11orf66; IIIG9; PPP1R32; SAXO4
UniProt ID: (Human) Q7Z5V6
Entrez Gene ID: (Human) 220004
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support