Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MLGRIKAQA FGFDQTFQSY RKDDFVMAFF KDPNVIPNLK LLSDSSGQWI ILGTEVKKIE AINVPCTQLS MSFFHRLYDE DIVRDSGHIV KCLDSFCDPF LISDELRRVL LVEDSEKYEI FSQPDREEFL FCLFKHLCLG GALCQYEDVI SPYLETTKLI YKDLVSVRKN PQTKKIQITS SVFKVSAYDS AGMCYPSAKN HEQTFSYFIV DPIRRHLHVL YHCYGVGDMS (39-267 aa encoded by BC006128)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cilia- and flagella-associated protein 300
Gene Aliases: C11orf70; CFAP300
UniProt ID: (Human) Q9BRQ4
Entrez Gene ID: (Human) 85016
Molecular Function: structural protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support