Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VIVTPGERR KQEMLKEMPL QDPRSREEAA RTQQLLLATL QEAATTQENV AWRKNWMVGG EGGASGRSP (26-94 aa encoded by BC090057 )
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Assembly factor CBP4 homolog; Ubiquinol-cytochrome-c reductase complex assembly factor 3; UPF0723 protein C11orf83
Gene Aliases: C11orf83; CCDS41658.1; MC3DN9; UNQ655; UNQ655/PRO1286; UQCC3
UniProt ID: (Human) Q6UW78
Entrez Gene ID: (Human) 790955
Molecular Function: chaperone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support