Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
This antibody is predicted to react with pig, sheep and cow based on sequence homology.
This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.
C12orf29 (Chromosome 12 Open Reading Frame 29) is a Protein Coding gene. Diseases associated with C12orf29 include Senior-Loken Syndrome 6 and Meckel Syndrome, Type 4. [GeneCards]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CLO29; DKFZp313K0436; DKFZp434N2030; DKFZp686L04169; FLJ38158; MGC102978; RNA ligase; RNA ligase 1; Rnl
Gene Aliases: C12orf29; RLIG1
UniProt ID: (Human) Q8N999
Entrez Gene ID: (Human) 91298
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support