Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Specificity: Human C12orf4
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ferry endosomal RAB5 effector complex subunit 3; Fy-3
Gene Aliases: C12orf4; FERRY3
UniProt ID: (Human) Q9NQ89
Entrez Gene ID: (Human) 57102
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support