Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LYIRSCRVLM LSDWYTMLYN PSPDYVTTVH CTH
Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
May be a regulator of the duration of MAPK8 activity in response to various stress stimuli. Facilitates degradation of misfolded endoplasmic reticulum (ER) luminal proteins through the recruitment of components of the proteasome and endoplasmic reticulum-associated degradation (ERAD) system.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: JAMP; JKAMP; JNK-associated membrane protein; JNK1-associated membrane protein; JNK1/MAPK8-associated membrane protein; Jun N-terminal kinase 1-associated membrane protein; Medulloblastoma antigen MU-MB-50.4
Gene Aliases: C14orf100; CDA06; HSPC213; HSPC327; JAMP; JKAMP
UniProt ID: (Human) Q9P055
Entrez Gene ID: (Human) 51528
Molecular Function: scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support