Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: AVHILTKLT ESSPYGPYTG QVLAVHILKA RKAYEHALQD CLGDSCVSNP APTDSCSRLI SLAKCFMLFQ YLTIGIDAAV QIYEQVFAKL NSSVFPEGSG EGDSASSQSW TSVLEAITLM HTSLLRFHMK VSVYPLAPLR EALSQALKLY PGNQVLWRSY VQIQNKSHSA SKTRRFFDTI TRSAKPLEPW LFAIEAEKLR KRLVETVQRL DGREIHATIP ETGLMHRIQA LFENAMRSDS GSQCPLLWRM YLNFLVSLGN KERSKGVFYK ALQSCPWAKV LYLDAVEYFP DEMQEILDLM TEKELRVRLP LEELELLLED (846-1164 aa encoded by BC118673)
NRDE2 gene ontology (GO) annotations include RNA interference; chromatin silencing by small RNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Nuclear exosome regulator NRDE2; Protein NRDE2 homolog; UPF0614 protein C14orf102
Gene Aliases: C14orf102; NRDE2
UniProt ID: (Human) Q9H7Z3
Entrez Gene ID: (Human) 55051
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support