Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Immunogen sequence: EVDEMEIEIN YLNQQISRHN ETKALSETLE EKNKNTENRK ELKERIFGKD EHVLTLNKTQ SSQLFLPYES QKLVRPIK
Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 56%.
Meiotic protein that localizes to the central element of the synaptonemal complex and is required for chromosome synapsis during meiotic recombination. Required for the appropriate processing of intermediate recombination nodules before crossover formation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Protein SIX6OS1; Six6 opposite strand transcript 1
Gene Aliases: C14orf39; SIX6OS1
UniProt ID: (Human) Q8N1H7
Entrez Gene ID: (Human) 317761
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support