Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotions
View available promotion(s)
Promo Code: CI2Z9E .
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
This gene encodes a member of a large family of mitochondrial transporters. The nuclear-encoded carrier protein is embedded in the inner mitochondrial membrane. This member of the family is thought to be an uncoupling protein that uncouples mitochondrial respiration from ATP synthesis by dissipating the transmembrane proton gradient. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HCC-down-regulated mitochondrial carrier protein; Hepatocellular carcinoma down-regulated mitochondrial carrier protein; hepatocellular carcinoma-downregulated mitochondrial carrier protein; Mitochondrial NAD(+) transporter SLC25A47; Solute carrier family 25 member 47; solute carrier family 25, member 47
Gene Aliases: C14orf68; HDMCP; HMFN1655; SLC25A47
UniProt ID: (Human) Q6Q0C1
Entrez Gene ID: (Human) 283600
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support