Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MRIEKCYFCS GPIYPGHGMM FVRNDCKVFR FCKSKCHKNF KKKRNPRKVR WTKAFRKAAG KELTVDNSFE FEKRRNEPIK YQRELWNKTI DAMKRVEEIK QKRQAKFIMN RLKKNKELQK VQDIKEVKQN IHLIRAPLAG KGKQLEEKMV
This gene encodes a protein sharing a low level of sequence similarity with human ribosomal protein L24. Although this gene has been referred to as RPL24, L30, and 60S ribosomal protein L30 isolog in the sequence databases, it is distinct from the human genes officially named RPL24 (which itself has been referred to as ribosomal protein L30) and RPL30. The protein encoded by this gene localizes to the nucleolus and is thought to play a role in the biogenesis of the 60S ribosomal subunit. The precise function of this gene is currently unknown. This gene utilizes alternative polyadenylation signals and has multiple pseudogenes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 60S ribosomal protein L30 isolog; homolog of yeast ribosomal like protein 24; my024 protein; Probable ribosome biogenesis protein RLP24; Ribosomal L24 domain-containing protein 1; Ribosomal protein L24-like
Gene Aliases: C15orf15; HRP-L30-iso; L30; My024; RLP24; RPL24; RPL24L; RSL24D1; TVAS3
UniProt ID: (Human) Q9UHA3
Entrez Gene ID: (Human) 51187
Molecular Function: ribosomal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support