Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
C15orf40 is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: UPF0235 protein C15orf40; UPF0235 protein C15orf40 homolog
Gene Aliases: AI851475; C15orf40
UniProt ID: (Mouse) Q9CRC3, (Human) Q8WUR7
Entrez Gene ID: (Mouse) 67290, (Human) 123207
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support