Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Sequence of this protein is as follows: MLDHKDLEAE IHPLKNEERK SQENLGNPSK NEDNVKSAPP QSRLSRCRAA AFFLSLFLCL FVVFVVSFVI PCPDRPASQR MWRIDYSAAV IYDFLAVDDI NGDRIQDVLF LYKNTNSSNN FSRSCVDEGF SSPCTFAAAV SGANGSTLWE RPVAQDVALV ECAVPQPRGS EAPSACILVG RPSSFIAVNL FTGETLWNHS SSFSGNASIL SPLLQVPDVD GDGAPDLLVL TQEREEVSGH LYSGSTGHQI GLRGSLGVDG ESGFLLHVTR TGAHYILFPC ASSLCGCSVK GLYEKVTGSG GPFKSDPHWE SMLNATTRRM LSHSSGAVRY LMHVPGNAGA DVLLVGSEAF VLLDGQELTP RWTPKAAHVL RKPIFGRYKP DTLAVAVENG TGTDRQILFL DLGTGAVLCS LALPSLPGGP LSASLPTADH RSAFFFWGLH ELGSTSETET GEARHSLYMF HPTLPRVLLE LANVSTHIVA FDAVLFEPSR HAAYILLTGP ADSEAPGLVS VIKHKVRDLV PSSRVVRLGE GGPDSDQAIR DRFSRLRYQS EA
ITFG3 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include cell surface; extracellular exosome; integral component of membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: family with sequence similarity 234, member A; integrin alpha FG-GAP repeat containing 3; Protein FAM234A; Protein ITFG3
Gene Aliases: C16orf9; FAM234A; gs19; ITFG3; RGD1311484
UniProt ID: (Human) Q9H0X4, (Rat) Q5M7W6
Entrez Gene ID: (Human) 83986, (Rat) 360502
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support