Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Peptide sequence: DLWRREAEAR KQSQPDPAMP PGHTRMPENQ RLETLTKLLQ SQSQLLRELV
Sequence homology: Cow: 86%; Dog: 86%; Guinea Pig: 92%; Horse: 85%; Human: 100%; Mouse: 83%; Rabbit: 77%; Rat: 92%
The exact function of C16orf48 remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cp048; Enkurin domain-containing protein 1
Gene Aliases: C16orf48; DAKV6410; ENKD1; UNQ6410/PRO21183
UniProt ID: (Human) Q9H0I2
Entrez Gene ID: (Human) 84080
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support