Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotions
View available promotion(s)
Promo Code: P5906365 Don't miss out: buy 3, only pay for 2 Extended to June 27: take advantage of our 3 for 2 promotion. Whether you are restocking essentials or trying something new, now is a great time to get more for your money!. Learn more
Promo Code: RPUZZ25 Stock up on essentials to piece your discovery together Until June 27, save up to $650 and get an exclusive lab-themed hidden-object puzzle. Learn more
Immunogen sequence: MLNRKTSHF LGMRVQSELE HLSELRREAG KDRSSVHGSA ARTRASVRTQ WTTAAAAKAD EDPGANLFPP PLPRPRICMW KYLDVHSMHQ LEKTTNAEMR EVLAELLELG CPEQSLRDAI TLDLFCHALI FCRQQGFSLE QTSAACALLQ DLHKACIATP LGNVEECYRY FTSVLFCHGV RRPPFSIDLF KEEQLLALED YVVNTYFRHF KLYKYVFTPQ VRLDLSLTYM GLQPPKLWPE SETEKEESKE MEEQAVTPQK EELETVAPPE PEPSHIHVLR AYIKTQVNKE LEQLQGLVEE RLKASEERLS SKLTALERPF QLPPGKGKSK TK (1-331 aa encoded by BC128129)
C16orf93, also named as CCDC189, is a coiled-coil domain-containing protein which has four isoforms with MW 38 kDa, 34 kDa, 12 kDa and 20 kDa.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cilia- and flagella-associated protein 119; Coiled-coil domain-containing protein 189
Gene Aliases: C16orf93; CCDC189; CFAP119
UniProt ID: (Human) A1A4V9
Entrez Gene ID: (Human) 90835
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support