Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAYEKSTDIS DVSRSMFLYP WLEYPDKTKE LRKAMAPVHL PLSCYQMPKE EFPPSPECWR QHPSKPNSVP YCYFKKPEIY THWHDLYDQR EEREAEKMLR KMRDDCRYIK EVHQTHIKMF HLPMSKLTIK SEMRSRPLEP TQDPLKWQRL RELTKSLESP REDEQFYAAQ ALGCLRISDK FVMEALQQVV SLRTAGVQER RKQGLRCVAC GHRRGSPGC
HEATR9 gene ontology annotations related to this gene include hematopoietic progenitor cell differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HEAT repeat-containing protein 9; Protein HEATR9
Gene Aliases: C17orf66; HEATR9
UniProt ID: (Human) A2RTY3
Entrez Gene ID: (Human) 256957
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support