Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAAPSGGWNG VGASLWAALL LGAVALRPAE AVSEPTTVAF DVRPGGVVHS FSHNVGPGDK YTCMFTYASQ GGTNEQWQMS LGTSEDHQHF TCTIWRPQGK SYLYFTQFKA EVRGAEIEYA MAYSKAAFER ESDVPLKTEE FEVTKTAVAH RPGAFKAELS KLVIVAKASR TEL
The interleukins are a broad family of well characterized cytokines, primarily of hematopoietic cell origin. Cytokines are small, soluble proteins with pleiotropic effects on a variety of cell types. Cytokines have a regulatory function over the immune system and mediate aspects of inflammatory response. IL-25 (also called SF20/IL-25) is a secreted bone marrow stroma-derived growth factor cytokine that is derived from Th2 T-cells. IL-25 belongs to the IL-17 cytokine family and is capable of amplifying allergic type inflammatory responses by its actions on other cell types. IL-25 binds to mouse thymic shared antigen-1 and supports lymphoid cell proliferation. Infusion of mice with IL-25 induces IL-4, IL-5, and IL-13 gene expression. IL-25 induced mice also develop epithelial cell hyperplasia, increased mucus secretion, and airway hyperreactivity, suggesting that IL-25 may be an important mediator of allergic disease via production of IL-4, IL-5, IL-13 and eotaxin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CS010; IL 25; IL 27; interleukin 25; interleukin 27 working designation; interleukin-25; Interleukin25; MYDGF; Myeloid-derived growth factor; R33729 1; R337291; stromal cell-derived growth factor SF20; UPF0556 protein C19orf10
Gene Aliases: C19orf10; EUROIMAGE1875335; IL25; IL27; IL27w; MYDGF; R33729_1; SF20
UniProt ID: (Human) Q969H8
Entrez Gene ID: (Human) 56005
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support