Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MWRGCARSRK RKKVDNDYNA LRERLSTLPD KLSYNIMVPF GPFAFMPGKL VHTNEVTVLL GDNWFAKCSA KQAVGLVEHR KEHVRKTIDD LKKVMKNFES RVEFTEDLQK MSDAAGDIVD IREEIKCDFE FKAKHRIAHK PHSKPKTSDI FEADIANDVK SKDLLADKEL WARLEELERQ EELLGELDSK PDTVIANGED TTSSEEEKED RNTNVNAMHQ VTDSHTPCHK DVASSEPFSG QVNSQLNCSV NGSSSYHSDD DDDDDDDDDD DNIDDDDGDN DHEALGVGDN SIPTIYFSHT VEPKRVRINT GKNTTLKFSE KKEEAKRKRK NSTGSGHSAQ ELPTIRTPAD IYRAFVDVVN GEYVPRKSIL KSRSRENSVC SDTSESSAAE FDDRRGVLRS ISCEEATCSD TSESILEEEP QENQKKLLPL SVTPEAFSGT VIEKEFVSPS LTPPPAIAHP ALPTIPERKE VLLEASEETG KRVSKFKAAR LQQKD
Involved in gene transcription regulation. Acts as a transcriptional repressor in concert with the corepressor UXT to regulate androgen receptor (AR) transcription. May act as a tumor suppressor to repress AR-mediated gene transcription and to inhibit anchorage-independent growth in prostate cancer cells. Required for cell survival in ovarian cancer cells. Together with UXT, associates with chromatin to the NKX3-1 promoter region. Antagonizes transcriptional modulation via hepatitis B virus X protein. Plays a central role in maintaining S6K1 signaling and BAD phosphorylation under normal growth conditions thereby protecting cells from potential deleterious effects of sustained S6K1 signaling. The URI1-PPP1CC complex acts as a central component of a negative feedback mechanism that counteracts excessive S6K1 survival signaling to BAD in response to growth factors. Mediates inhibition of PPP1CC phosphatase activity in mitochondria. Coordinates the regulation of nutrient-sensitive gene expression availability in a mTOR-dependent manner. Seems to be a scaffolding protein able to assemble a prefoldin-like complex that contains PFDs and proteins with roles in transcription and ubiquitination. [Uniprot]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ10575; Protein NNX3; Protein phosphatase 1 regulatory subunit 19; protein phosphatase 1, regulatory subunit 19; RNA polymerase II subunit 5-mediating protein; RPB5-mediating protein; Unconventional prefoldin RPB5 interactor 1; URI1 prefoldin-like chaperone
Gene Aliases: C19orf2; NNX3; PPP1R19; RMP; URI; URI1
UniProt ID: (Human) O94763
Entrez Gene ID: (Human) 8725
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support