Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: ETFHPFVPEH HLIKGYLPRT FWYWNYNYYP PVEGPGCCSD LAVSFHYVDS TTMYELEYLV YHLRPYGYLY RYQPTLPERI LKEISQANKN EDTKVKLGNP
Glycosyltransferase that generates the core 1 O-glycan Gal-beta1-3GalNAc-alpha1-Ser/Thr (T antigen), which is a precursor for many extended O-glycans in glycoproteins. Plays a central role in many processes, such as angiogenesis, thrombopoiesis and kidney homeostasis development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B3Gal-T8; Beta-1,3-galactosyltransferase; C1GalT1; Core 1 beta1,3-galactosyltransferase 1; core 1 beta3-Gal-T1; Core 1 O-glycan T-synthase; Core 1 synthase; core 1 synthase, glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase, 1; Core 1 UDP-galactose:N-acetylgalactosamine-alpha-R beta 1,3-galactosyltransferase 1; Glycoprotein-N-acetylgalactosamine 3-beta-galactosyltransferase 1
Gene Aliases: C1GALT; C1GALT1; T-synthase
UniProt ID: (Human) Q9BTU1
Entrez Gene ID: (Human) 56913
Molecular Function: transferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support